Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wrt-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076196.7 | NP_508597.2 | 2 - 3 | 213 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Caenorhabditis elegans Probe spans exons |