Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ncr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076370.5 | NP_508771.1 | 17 - 18 | 4164 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |