Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029194.4 | NP_001024365.1 | 7 - 8 | 1067 | 96 | |
NM_001380949.1 | NP_001367445.1 | 8 - 9 | 992 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |