Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hyl-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076402.9 | NP_508803.3 | 3 - 4 | 314 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Caenorhabditis elegans Probe spans exons |