Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ztf-16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029679.5 | NP_001024850.1 | 8 - 9 | 946 | 122 | |
NM_001029680.5 | NP_001024851.1 | 8 - 9 | 952 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |