Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pqn-62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313445.3 | NP_001300374.1 | 14 - 15 | 3444 | 109 | |
NM_001313446.4 | NP_001300375.1 | 16 - 17 | 3741 | 109 | ||
NM_001380960.1 | NP_001368582.1 | 16 - 17 | 3720 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |