Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sto-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270091.4 | NP_001257020.1 | 3 - 4 | 290 | 83 | |
NM_001270092.5 | NP_001257021.1 | 3 - 4 | 2001 | 83 | ||
NM_001270093.4 | NP_001257022.1 | 3 - 4 | 283 | 83 | ||
NM_001270094.2 | NP_001257023.1 | 2 - 3 | 174 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |