Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: jkk-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383579.1 | 3 - 4 | 425 | 63 | ||
GenBank mRNA | AB024086.1 | - | 3 - 4 | 365 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |