Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-97 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076542.8 | NP_508943.3 | 1 - 2 | 169 | 60 | |
GenBank mRNA | AF035583.1 | - | 1 - 2 | 171 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |