Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tbc-16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076587.7 | NP_508988.3 | 16 - 17 | 2190 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |