Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nnt-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076627.6 | NP_509028.1 | 16 - 17 | 2778 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |