Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: irk-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076737.5 | NP_509138.2 | 4 - 5 | 547 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |