Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cah-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076785.5 | NP_509186.3 | 3 - 4 | 317 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Caenorhabditis elegans Probe spans exons |