Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: clc-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001375068.3 | NP_001362078.1 | 2 - 3 | 162 | 123 | |
NM_076856.6 | NP_509257.1 | 2 - 3 | 155 | 123 | ||
GenBank mRNA | AF252604.1 | - | 2 - 3 | 157 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |