Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: coh-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001322622.4 | NP_001309700.1 | 2 - 3 | 215 | 78 | |
NM_001322623.4 | NP_001309701.1 | 1 - 2 | 134 | 78 | ||
NM_076861.7 | NP_509262.2 | 7 - 8 | 1522 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Caenorhabditis elegans Probe spans exons |