Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cutl-29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076994.5 | NP_509395.2 | 2 - 3 | 160 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |