Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ugt-23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083286.4 | NP_001076755.1 | 7 - 8 | 1492 | 140 | |
NM_001083287.6 | NP_001076756.1 | 6 - 7 | 1122 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |