Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nas-37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029242.6 | NP_001024413.1 | 13 - 14 | 1841 | 67 | |
NM_001029243.6 | NP_001024414.1 | 13 - 14 | 1600 | 67 | ||
GenBank mRNA | AY912076.1 | - | 13 - 14 | 1615 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |