Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nas-37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029242.6 | NP_001024413.1 | 1 - 2 | 76 | 73 | |
NM_001029243.6 | NP_001024414.1 | 1 - 2 | 75 | 73 | ||
GenBank mRNA | AY912076.1 | - | 1 - 2 | 90 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |