Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gnrr-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077284.4 | NP_509685.2 | 3 - 4 | 366 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |