Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: moc-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077299.6 | NP_509700.2 | 9 - 10 | 1985 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |