Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: oac-14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313409.4 | NP_001300338.1 | 2 | 850 | 140 | |
NM_077300.6 | NP_509701.1 | 3 - 4 | 1138 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |