Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: abts-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077535.7 | NP_509936.3 | 8 - 9 | 994 | 65 | |
GenBank mRNA | AY887904.1 | - | 8 - 9 | 1016 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |