Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cab-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077635.6 | NP_510036.1 | 8 - 9 | 1268 | 79 | |
GenBank mRNA | AF293976.1 | - | 6 - 7 | 1081 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Caenorhabditis elegans Probe spans exons |