Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: str-10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001322638.1 | NP_001309672.1 | 5 - 6 | 946 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |