Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: twk-23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270300.3 | NP_001257229.1 | 8 - 9 | 951 | 79 | |
NM_001270301.4 | NP_001257230.1 | 7 - 8 | 855 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |