Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: elt-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270307.4 | NP_001257236.1 | 2 - 3 | 221 | 73 | |
NM_001270308.4 | NP_001257237.1 | 1 - 2 | 104 | 73 | ||
NM_001381138.1 | NP_001367208.1 | 5 - 6 | 548 | 73 | ||
NM_001381139.1 | NP_001366766.1 | 6 - 7 | 650 | 73 | ||
GenBank mRNA | AF146517.1 | - | 3 - 4 | 358 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |