Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: del-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001368544.2 | NP_001355359.1 | 2 - 3 | 254 | 103 | |
NM_001368545.3 | NP_001355470.1 | 3 | 398 | 103 | ||
NM_001368546.2 | NP_001355471.1 | 2 - 3 | 203 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |