Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vit-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076467.6 | NP_508868.1 | 4 - 5 | 4599 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |