Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srx-80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028744.1 | NP_001023915.1 | 5 - 6 | 750 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |