Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: adpr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026658.2 | NP_001021829.1 | 1 - 2 | 117 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |