Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nas-23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027110.3 | NP_001022281.1 | 6 - 7 | 1063 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |