Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sre-34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027224.1 | NP_001022395.1 | 4 - 5 | 884 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |