Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-69 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027581.5 | NP_001022752.1 | 2 - 3 | 332 | 109 | |
NM_001027582.4 | NP_001022753.1 | 2 - 3 | 332 | 109 | ||
NM_001129245.4 | NP_001122717.1 | 2 - 3 | 269 | 109 | ||
GenBank mRNA | AY919833.1 | - | 2 - 3 | 272 | 109 |
Gene Symbol: sas-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027583.4 | NP_001022754.1 | 2 - 3 | 265 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |