Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ttr-38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047783.3 | NP_001041248.1 | 1 - 2 | 156 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |