Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BMP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001287170.1 | NP_001274099.1 | 2 - 3 | 329 | 84 | |
XM_022421760.1 | XP_022277468.1 | 4 - 5 | 494 | 84 | ||
XM_022421761.1 | XP_022277469.1 | 4 - 5 | 430 | 84 | ||
XM_022421762.1 | XP_022277470.1 | 5 - 6 | 628 | 84 | ||
XM_022421763.1 | XP_022277471.1 | 4 - 5 | 417 | 84 | ||
XM_022421764.1 | XP_022277472.1 | 6 - 7 | 667 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris Probe spans exons |