Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHOB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001287052.1 | NP_001273981.1 | 1 | 955 | 58 | |
GenBank mRNA | AJ388536.1 | - | 1 | 109 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Inventoried Both primers and probe map within a single exon |