Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCER1G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003171.2 | NP_001003171.2 | 2 - 3 | 145 | 76 | |
XM_014111133.2 | XP_013966608.1 | 3 - 4 | 537 | 76 | ||
GenBank mRNA | AB017553.1 | - | 2 - 3 | 145 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Canis familiaris Probe spans exons |