Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRD5A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001287100.1 | NP_001274029.1 | 728 | 102 | ||
XM_022414068.1 | XP_022269776.1 | 609 | 102 | |||
XM_022414070.1 | XP_022269778.1 | 1238 | 102 | |||
XM_022414071.1 | XP_022269779.1 | 2245 | 102 | |||
GenBank mRNA | AF136720.1 | - | 54 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Canis familiaris |