Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003077.1 | NP_001003077.1 | 454 | 64 | ||
GenBank mRNA | AF328930.1 | - | 318 | 64 | ||
EU426968.1 | - | 447 | 64 | |||
U33843.1 | - | 454 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Canis familiaris |