Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FMO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003060.2 | NP_001003060.1 | 8 - 9 | 1372 | 141 | |
GenBank mRNA | AF384054.1 | - | 7 - 8 | 1328 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Canis familiaris Inventoried Probe spans exons |