Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASB17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003018.2 | NP_001003018.2 | 2 - 3 | 789 | 96 | |
GenBank mRNA | AJ518867.1 | - | 2 - 3 | 796 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Canis familiaris Probe spans exons |