Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NGF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194950.1 | NP_001181879.1 | 1 | 201 | 76 | |
GenBank mRNA | AJ626845.1 | - | 1 | 49 | 76 | |
AY800386.1 | - | 1 | 82 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Canis familiaris Inventoried Both primers and probe map within a single exon |