Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCNB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005763.1 | NP_001005763.1 | 4 - 5 | 483 | 139 | |
XM_022415500.1 | XP_022271208.1 | 4 - 5 | 477 | 139 | ||
XM_022415501.1 | XP_022271209.1 | 4 - 5 | 477 | 139 | ||
XM_022415502.1 | XP_022271210.1 | 4 - 5 | 477 | 139 | ||
XM_022415503.1 | XP_022271211.1 | 4 - 5 | 514 | 139 | ||
XM_022415504.1 | XP_022271212.1 | 4 - 5 | 481 | 139 | ||
XM_022415505.1 | XP_022271213.1 | 4 - 5 | 482 | 139 | ||
GenBank mRNA | AJ833648.1 | - | 4 - 5 | 483 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |