Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANP32A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003013.2 | NP_001003013.2 | 5 - 6 | 622 | 66 | |
GenBank mRNA | AY162293.1 | - | 5 - 6 | 622 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Canis familiaris Probe spans exons |