Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DEFB122 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_132650.1 | 3 - 4 | 398 | 93 | ||
GenBank mRNA | AY169791.1 | - | 3 - 4 | 321 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Canis familiaris Probe spans exons |