Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HPRT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003357.2 | NP_001003357.2 | 2 - 3 | 283 | 96 | |
GenBank mRNA | AY283372.1 | - | 2 - 3 | 134 | 96 | |
DN394227.1 | - | 2 - 3 | 293 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Canis familiaris Probe spans exons |