Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HPRT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003357.2 | NP_001003357.2 | 6 - 7 | 633 | 102 | |
NM_001313818.1 | NP_001300747.1 | 5 - 6 | 526 | 102 | ||
GenBank mRNA | AY283372.1 | - | 6 - 7 | 484 | 102 | |
DN394227.1 | - | 6 - 7 | 643 | 102 | ||
DN418028.1 | - | 5 - 6 | 512 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Inventoried Probe spans exons |