Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003226.2 | NP_001003226.1 | 5 - 6 | 619 | 135 | |
XM_005617216.3 | XP_005617273.1 | 5 - 6 | 694 | 135 | ||
XM_005617217.3 | XP_005617274.1 | 5 - 6 | 694 | 135 | ||
XM_022405358.1 | XP_022261066.1 | 5 - 6 | 664 | 135 | ||
GenBank mRNA | AY289762.1 | - | 5 - 6 | 605 | 135 | |
U50746.1 | - | 5 - 6 | 620 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |