Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHCE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001048036.2 | NP_001041501.1 | 3 - 4 | 553 | 75 | |
GenBank mRNA | AY831676.1 | - | 3 - 4 | 553 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris Probe spans exons |